Sodium chloride in aqueous solution USP Test Solution (TS)
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85372.180EA
82.8
GBP
85372.180
85372.260
Sodium chloride in aqueous solution USP Test Solution (TS)
Sodium chloride
Ready to use test solutions (TS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
Formula:
NaCl MW: 58.44 g/mol Melting Pt: 801 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 0 Merck Index: 14,08599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...