Potassium sulphate in aqueous solution USP Test Solution (TS)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
85368.260EA
35
GBP
85368.260
85368.180
Potassium sulphate in aqueous solution USP Test Solution (TS)
Potassium sulphate
Ready to use test solutions (TS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
Formula:
K₂SO₄ MW: 174.26 g/mol Melting Pt: 1069 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 Merck Index: 12,07845 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...