Would you like to log in again?
Transfer buffer, 10X Western Blot Transfer buffer, Pierce™
Supplier: Thermo Scientific
This transfer buffer is a methanol-free, space saving, stock solution for the preparation of methanol-free transfer buffer that is used with cooled, tank (wet) Western blotting transfer apparatus.
  • Easy to use - no packets to open, no powder to dissolve, and no methanol required
  • Increased accuracy
  • Saves time and space
For research use only. Not for use in diagnostic procedures.
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Pierce™ 10X Western blot transfer buffer, methanol-free 5 l PIER35040
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty