Azelaic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,9-Nonanedioic acid1,7-Heptanedicarboxylic acid

SPCMA2215-5KGBLEA 5410 GBP
SPCMA2215-5KGBL SPCMA2215-25KGBL SPCMA2215-1KGBL
Azelaic acid
Azelaic acid
Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 106 °C
Density: 1.0287 g/cm³ (20 °C)
Flash Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00004432
CAS Number: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908

Order Now

Specification Test Results

Assay 85% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR