3-Fluoro-2-nitrobenzoic acid 98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H63051.MDEA
19.6
GBP
H63051.MD
H63051.06
3-Fluoro-2-nitrobenzoic acid 98%
3-Fluoro-2-nitrobenzoic acid
Formula:
C₇H₄FNO₄ MW: 185.11 g/mol |
MDL Number:
MFCD07368342 CAS Number: 1000339-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...