Palladium 0.5% on alumina, spheres 2-4 mm

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

41383.14EA 78.8 GBP
41383.14 41383.22
Palladium 0.5% on alumina, spheres 2-4 mm
Palladium

Applications: Gas purification (O2, O3)

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR