Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sun protection cream, Stokoderm® Sun Protect 30 PURE

Supplier: Deb Stoko
Stokoderm® Sun Protect 30 PURE
Perfume-free, water resistant 5-star rated, factor 30 sunscreen for professional use to protect the skin against UVA, UVB and UVC exposure. For use on any part of the body exposed to the sun whilst working outdoors. Suitable for all skin types.

  • Provides 30 times the skin’s normal protection against the sun’s UVB rays
  • Water resistant formula withstands heavy perspiration or immersion in water; ideal for outdoor workers
  • Contains vitamin E, moisturisers and an emollient matrix of jojoba oil, cocoa butter, caprylic/capric triglyceride and glycerine to hydrate the skin
  • No greasy film, absorbs quickly
  • pH-neutral, colourant-free, perfume-free




 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Sun protection creams, Stokoderm® Sun Protect 30 PURE, 1000 ml cartridge 6 115-2374
Sun protection creams, Stokoderm® Sun Protect 30 PURE, 100 ml tube 12 115-2088
Sun protection creams, Stokoderm® Sun Protect 30 PURE, 30 ml tube 30 115-2375
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Colour Volume Pk VWR Catalog Number Quantity
Sun cream dispenser, Sun Protect 1000 White 1 l 1 EVONSUN1LDSMD
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping