Gel for optimal hand disinfection without water with simultaneous skin care by means of a moisturising complex. Effective against bacteria (including MRSA), fungi and enveloped viruses. For use without water.
- Effective against bacteria(incl. MRSA), fungi
- Bacterial reduction: 99.999 %
- Tuberculocid in accordance with EN 14348
- Virucidal against enveloped viruses, effective against A/H1N1 virus in line with EN 14476
- Contains glycerine and panthenol
- Alcohol based but kind to the skin
- Fragrance-free
Certifications: EN 1500, EN 12791, EN 13727, EN 1276, EN 1650, HACCP