Would you like to log in again?
Filter tips for Rainin LTS style pipettes, xTIP®
Supplier: Biotix
FlexFit®, X-Resin®, and Blade® technologies developed by Biotix are found exclusively in Biotix and xTIP brand pipette tips. Biotix technologies minimise inaccuracies associated with the number one issue in pipetting: user error. These proven features are standard on all xTIP Low Retention pipette tips, maximising accuracy and precision.
  • Perfect fit thanks to FlexFit technology: FlexFit’s flexible proximal end ensures confidence in a secure seal
  • Clean delivery with X-Resin and Blade technologies
  • X-Resin is a sleek resin that needs no coating, resulting in uniform composition throughout xTIP from proximal end to distal end
  • X-Resin provides ultra-low retentive qualities, with every lot, from the first tip to the last
  • Blade technology eliminates hanging droplet formation, ensuring sample precision with each and every delivery by reducing surface tension at the distal end of the tip
  • Certified free of RNase, DNase, pyrogens, PCR inhibitors
Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for precision, ensuring superior quality and compatibility. All raw materials are traceable.
Each tray includes a label with a unique bar-coded serial number, part number, and description for specific traceability.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Tip style Volume Sterile Filter Tip colour For use with Packed Pk VWR Catalog Number Quantity
Low retention (X-Resin) 20 µl + + Transparent Rainin LTS style pipettes 10 racks, 96 each 960 732-2909
Low retention (X-Resin) 200 µl + + Transparent Rainin LTS style pipettes 10 racks, 96 each 960 732-2911
Low retention (X-Resin) 1000 µl + + Transparent Rainin LTS style pipettes 10 racks, 96 each 960 732-2912
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty