Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Digital camera, VisiCam® HRS 14

Supplier: VWR Collection
VWR® VisiCam®
CMOS camera with high speed output, built-in buffer 32 Mb memory and image transmission.

  • C-mount attachment (not included) to be attached to trinocular microscopes or to binocular/monocular microscopes through 0,45× eyepiece adapter
  • Twain and Directshow compatible software
  • Measurement functions, calibration, scale marker, annotations and time-lapse

It is recommended for 4K (Ultra High Definition) applications, which require high frame rate and image quality, like semiconductor and materials inspection, defect analysis metrology, quality inspection, and life science applications.

Ordering information: Supplied with USB cable and multi-language interface software, interactive tools customised by user using macro or advanced measuring tools (calibration with Interactive Scale Marker can be saved protecting any unexpected changes).




 

ORDER

Expand All / Collapse All
Description Model Resolution Pk VWR Catalog Number Quantity
High resolution digital camera HRS 14 14 MP 1 630-2671

Adapters
Description Pk VWR Catalog Number Quantity
Eyepiece adapter, 0,45×, Ø 23/30 mm 1 630-2674
CCD camera adapter, 1/2" sensor, for stereo microscope 300 series 1 630-1793
CCD camera adapter, 1/2" sensor, for 500 series 1 630-1749
- 1 630-1892 Unavailable
CCD camera adapter for IT 404 1 630-1768
unavailable This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Max. resolution 14 MP
Frame rate 12 fps @ 14 MP
Interfaces USB 3.0
Supported OS Windows XP, 7, 8, 10
W×D×H 68×45×35 mm
Weight 160 g

ACCESSORIES

Expand All / Collapse All

Accessories
Description Pk VWR Catalog Number Quantity
Calibration slide with circles and ruler 1 630-1123
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty