Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra-low temperature upright freezers, −40 °C

Supplier: VWR Collection
These upright freezers have been designed for sample protection and ease of use.

  • Heavy gauge, cold-rolled steel cabinet construction with a powder coat paint finish, resisting chipping and rust
  • Four inner doors reduce cold air loss and improve temperature recovery after door openings
  • Insulation: high density HFC blown PU foam (127 mm)
  • Sturdy, stainless steel shelving (adjustable in 25 mm increment, max. capacity: 45,4 kg)
  • Gaskets maximise cabinet temperature and work to eliminate frost build-up
  • Single handed operation with ergonomic, padlock compatible door handle featuring an integrated key lock
  • 2” locking castors for easy to roll installation
  • Bright, easy to read temperature display shows actual and (optional) sample probe temperatures
  • Refrigerant type: R 404-A
  • Information controller for simplified control and monitoring
  • RS485 interface
  • Optional temperature chart recorder (7-day) and optional data logger can be connected

Temperature range: −10 to −40 °C

Certifications: Regulatory certifications: UL, cUL, CE




 

ORDER

Expand All / Collapse All
Type Pk VWR Catalog Number Quantity
ULT freezer, 368 l 1 471-1135
ULT freezer, 490 l 1 471-1136
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model 471-1135 471-1136
Capacity (l) 368 490
Temperature (°C) −10...−40
Display Digital
Defrost Manual
Alarm audible and visual
Shelves/drawers 4 shelves/4 compartments
Doors 1 outer (right hinge) / 4 inner
Door lock Yes
Material Exterior: Cold rolled steel, powder coated
Interior: Galvaneal
Specifications Storage capacity for 2" boxes: 240
Storage capacity for 3" boxes: 144
Storage capacity for vials: 24000
Storage capacity for 2" boxes: 320
Storage capacity for 3" boxes: 192
Storage capacity for vials: 32000
Access ports 2
W×D×H int. (mm) 572×492×1308 572×665×1308
W×D×H ext. (mm) 912×802×1978 912×988×1978
Weight (kg) 265 345
Noise level 61 dB(A)
Electrical 230 V, 50 Hz, 1 phase, 3450 W

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty