Anti-CSP1 Rabbit Polyclonal Antibody
BOSSBS-11541R
: Bioss
- Antibody type:Primary
- Antigen name:Calcipressin 1
- Antigen symbol:CSP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Antigen synonyms:Down syndrome critical region gene 1|Calcium and oxidant inducible mRNA|DSCR1|Down syndrome candidate region 1|Calcipressin 1|Modulatory calcineurin interacting protein 1|RCN 1|DSCR-1|Adapt 78|Myocyte enriched calcineurin interacting protein 1|Near DSCR proline rich protein|DSCR 1|Regulator of calcineurin 1.|MCIP 1|RCAN1|DSC1|Calcipressin-1|RCN1|Myocyte-enriched calcineurin-interacting protein 1|CSP 1|Adapt78|CSP1|MCIP1|DSC 1|Down syndrome critical region protein 1|RCAN1_HUMAN
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
DSCR1 protein influences cardiac and nervous system development (1). DSCR1 is abundant in the brain, heart, and skeletal muscle. Overexpression of DSCR1 may play a role in the pathogenesis of Down syndrome, in particular mental retardation and/or cardiac defects. DSCR1 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A. Human DSCR1 maps to gene locus 21q22.1-q22.2.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: CSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: