United Kingdom
Order Entry
Anti-WSB1 Rabbit Polyclonal Antibody
  ABNOPAB29394
 :  Abnova
Anti-WSB1 Rabbit Polyclonal Antibody
  ABNOPAB29394
 :  Abnova
 
£517.00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    WD repeat and SOCS box containing 1
  • Antigen symbol:
    WSB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    26118
  • Antigen synonyms:
    WSB-1|SWIP1
  • Sequence:
    APFDKKCGRENWTVAFAPDGSYFAWSQGHRTVKLVPWSQCLQNFLLHGTKNVTNSSSLRLPRQNSDGGQKNKPREHIIDCGDIVWSLAFGSSVPEKQSRCVNIEWHRFRFGQDQLLLATGLNNGRIKIWDV
  • Immunogen:
    Recombinant protein corresponding to human WSB1.
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against recombinant human WSB1.

Type: Primary
Antigen: WSB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human