Half mask respirator, FM1
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SCTS2023539EA 68.2 GBP
SCTS2023539 SCTS2023538
Half mask respirator, FM1
The reusable half mask FM1 is designed with liquid silicone rubber face seal for exceptional comfort and fit. The low profile design of the mask makes it compatible with visors, welding shields and other PPE. Ideal for welding and spraying.
  • Integrates easily with other PPE
  • Side entry hose improves comfort and balance
  • Compatible with Spirit, Tornado, Proflow2 120SC/120EX, 160SC/160EX & Proflow3 120EX
Certifications: EN12942 TM2
Ordering information: FM1 is supplied without hose and that should be ordered separately.