Would you like to log in again?
Bump cap, First Base 3 Flame Retardant
Supplier: Scott Safety
First Base 3 Flame Retardant
The First Base 3 flame-retardant bump protection cap is designed to protect workers where they are exposed to flammable liquids or hot sparks that could potentially burn the head or ignite hair.
  • No direct ventilation holes minimises potential burn risk
  • Completely flame-retardant (coated cotton) outer cap
  • High performance sweatband absorbs and wicks moisture away quickly
  • Flexible ABS protective shell guards against bumps and lacerations to the head
Colour: Navy blue
Certifications: EN 812:A1
ORDER
Expand All / Collapse All
Description Colour Pk VWR Catalog Number Quantity
Fire retardant, reduced peak Dark blue 1 111-2260
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty