You Searched For: Paint/Ink Film Applicators

Multi-functional applicators are suitable for applying paint films of various different predefined thicknesses. Automated film applicators provide a reliable basis to apply coating films and slurries to test charts, panels, or foils in a uniform and reproducible way. Prism applicators are suitable for applying a host of different products onto flat and relatively solid substrates. Models constructed of high-grade stainless steel are resilient against acid or base elements.

Multi-functional applicators are suitable for applying paint films of various different predefined thicknesses. Automated film applicators provide a reliable basis to apply coating films and slurries to test charts, panels, or foils in a uniform and reproducible way. Prism applicators are suitable for applying a host of different products onto flat and relatively solid substrates. Models constructed of high-grade stainless steel are resilient against acid or base elements.


9  results were found

SearchResultCount:"9"

Sort Results

List View Easy View

Rate These Search Results

Supplier: Industrial Physics
Description: Spiral type applicator with a film width of 250 mm and available in ranges from 4 to 200 μm. Ideal for application of film onto thin materials, such a...

Supplier: Industrial Physics
Description: Multi-functional film applicator with 4 application sides for applying paint films of 4 different predefined thicknesses. The applicators are availabl...

Supplier: Industrial Physics
Description: Cylindrical film applicator with four application sides for applying paint films of four different predefined thicknesses in film width 60 or 80 mm. S...

Supplier: Industrial Physics
Description: Bird-type film applicators feature one flat edge and one application side for applying a pre-defined coating thickness. Bird-type applicator single ga...

Catalog Number: (SIMEVF1590)
Supplier: Industrial Physics
Description: Ideal for applying paint with use of a automatic film applicator or drying time recorder.
UOM: 1 * 1 items


Supplier: Industrial Physics
Description: Stainless steel spiral applicators with a film width of 320 mm are round bars tightly wound with stainless steel wire. The diameter of the wire, in wo...

Supplier: Industrial Physics
Description: Prismatic film applicator with four application sides for applying paint films of four different predefined thicknesses. The prism applicators are ava...

Supplier: Industrial Physics
Description: The Automatic Film Applicator Standard provides a reliable basis to apply coating films to test charts, panels or foils in a uniform and reproducible ...

Catalog Number: (SIMEVF1550)
Supplier: Industrial Physics
Description: Multi-functional film applicator (Octoplex) with 8 pre-defined thickness sides for applying paint films of 25, 50, 75, 100, 125, 150, 175, 200 µm. Fil...
UOM: 1 * 1 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on 0800 22 33 44
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
Additional Documentation may be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
1 - 9 of 9
no targeter for Bottom