Pressure lever cassettes
Supplier: Ismatec
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ISMCIS3820AEA 213 GBP
ISMCIS3820A
Pressure lever cassettes
This optional pressure lever allows to set different tubing pressure for each channel. Depend on the tubing material and diameter an optimally adjusted tubing pressure can be set. To maintain constant flow rates it may be necessary to periodically adjust the tubing pressure.