Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cross cut adhesion test kits, CC1000

Supplier: TQC B.V.
This Cross Cut adhesion test kit (CC1000) is used to test the adhesion of dry coats of paint on their substrate by means of a series of cuts through the coating.

  • Round cutting knife can be changed easily by rotating the knife
  • Self-adjusting knife-holder ensures equal pressure on the cutting knife
  • Round cutting knife with eight cutting edges for longer lifetime
  • Ergonomically shaped handle
  • Wide range of knife sizes available for different coating thicknesses and substrates and according to different standards

Two series of parallel cuts cross angled to each other to obtain a pattern of 25 or 100 similar squares. The ruled area is evaluated by using a table chart after a short treatment with a stiff brush, or adhesive tape for hard substrates.

Certifications: EN ISO 2409, ASTM D3359

Delivery information: Kit includes soft grip handle incl. CC1000 holder, adhesive tape (adhesion to steel 4,3 N/cm), hardened steel cutter (type may vary, see below), 4 mm Allen key, nylon brush, sturdy case and illuminated magnifier.




 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Cross Cut adhesion test CC1000, 6 blades, cutting distance 1 mm, acc. to ISO 2409 & ASTM D3359 1 SIMEVF1839
Cross Cut adhesion test CC1000, 6 blades, cutting distance 2 mm, acc. to ISO 2409 & ASTM D3359 1 SIMEVF1842
Cross Cut adhesion test CC1000, 6 blades, cutting distance 3 mm, acc. to ISO 2409 1 SIMEVF1844
Cross Cut adhesion test CC1000, 11 blades, cutting distance 1 mm, acc. to ASTM D3359 <2009 1 SIMEVF1846
Cross Cut adhesion test CC1000, 11 blades, cutting distance 1,5 mm, acc. to ASTM D3359 <2009 1 SIMEVF1847
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Width Pk VWR Catalog Number Quantity
Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003 25,0 mm 1 SIMESP3007
Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003 25,0 mm 3 Roll SIMESP3010
Spare knife, round, 1 mm, acc. to ISO 2409 & ASTM D3359, suitable for C1000 test kits - 1 SIMEVF2355
Spare knife, round, 1 mm, acc. to ASTM D3359 <2009, suitable for C1000 test kits - 1 SIMEVF2359
Spare knife, round, 1,5 mm acc. to ASTM D3359 <2009, suitable for C1000 test kits - 1 SIMEVF1861
Spare knife, round, 2 mm, acc. to ISO 2409 & ASTM D3359, suitable for C1000 test kits - 1 SIMEVF2357
Spare knife, round, 3 mm, acc. to ISO 2409, suitable for C1000 test kits - 1 SIMEVF2358
Nylon brush for cross cut adhesion test - 1 SIMESP1710
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping