- Antibody type:Primary
- Antigen name:osm-9-like TRP channel 2
- Antigen symbol:VRL-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:at VRL-1 protein (aa: 744-761)
- Cross adsorption:No
- Format:Lyophilized
- Immunogen:A synthetic peptide (C-KNSASEEDHLPLQVLQSP) as part of rat VRL-1 protein (aa: 744-761) conjugated to KLH has been used as the immunogen.
- Size:50 µg
- Pk:50 µG
TISSUE SPECIFICITY: Ubiquitously expressed. Expressed in dorsal root ganglia, trigeminal ganglia, spinal chord (Lissauer's tract, dorsal horn and dorsal columns) (at protein level). PTM: N-glycosylated. PTM: Phosphorylated by PKA. SIMILARITY: Belongs to the transient receptor family. TrpV subfamily. SIMILARITY: Contains 3 ANK repeats.
Immunohistochemical analysis in rat dorsal root ganglia and spinal cord indicates a high level of specificity for this antiserum. Specificity was also shown by Western blot.
Application Information:
IHC, Immunofluorescence, Western blot and Immunoblot. Recommended to be used at a concentration of 0.5-1 µg for these applications. This antiserum works superbly for immunohistochemistry on free-floating or paraffin embedded sections. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: VRL-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: at VRL-1 protein (aa: 744-761)
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human