Disinfectant wipes, Premier®
Supplier: Bunzl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Premier®
BUNZPM7200EA 106 GBP
BUNZPM7200
Disinfectant wipes, Premier®
Wipes Disposable Wipes and Towels Disinfectant Wipes
These disinfectant wipes are designed for use on all hard surfaces to provide effective protection against E coli, staphylococcus aureus, pseudomonas aeruginosa, streptococcus faecalis, salmonella typhimurium, listeria monocytogenes, candida albican and saccharomyces cerevisiae.
  • White, non woven material
  • Impregnated with 70% Ethanol solution for fast, effective decontamination
  • Perforated for easy dispensing
Colour: White
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904