Anti-HIS Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOPAB29062EA 556 GBP
ABNOPAB29062
Anti-HIS Polyclonal Antibody
Antibodies
Chicken polyclonal antibody raised against recombinant HIS peptide.

Recommended Dilutions: ChIP: 1:5000-1:10000; Immunoprecipitation:1:5000-1:10000

Type: Primary
Antigen: HIS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR