Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Supplier: Abnova
Chicken polyclonal antibody raised against recombinant HA peptide.

Recommended Dilutions: Immunocytochemistry Immunohistochemistry: 1:250; Western Blot

Type:
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host:
Isotype: IgY
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
1 mL 1 ml ABNOPAB29058
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol HA
Antigen name Hemagglutinin
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Isotype IgY
Western blot Yes
ImmunoChemistry Yes
Format Liquid
Cross adsorption No
Immunogen A KLH-conjugated synthetic peptide corresponding to HA tag emulsified in Freund's adjuvant.
Purification Antigen affinity purification
Sequence YPYDVPDYA
Storage buffer PBS, pH 7,2 (0,2% BSA, 0,02% sodium azide)
Storage temperature Store at 4 °C and avoid from light. For long term storage, dilute the antibody with sterile glycerol and store it at – 20 °C. Aliquot to avoid repeated freezing and thawing.

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty