Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC12A1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: Abnova
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.

Recommended Dilutions: Western Blot: 1:1000

Type:
Antigen: SLC12A1
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 μg 100 µg ABNOPAB29045
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol Solute carrier family 12, member 1
Antigen name Solute Carrier Family 12 Member A1
Conjugation AP (Alkaline Phosphatase)
Clonality Polyclonal
Host Rabbit
Western blot Yes
Antigen synonyms NKCC2, Nkcc2, BSC1
Amino acid number 33 to 55
Gene ID 25065
Epitope N-Terminal
Format Liquid
Cross adsorption No
Immunogen A synthetic peptide corresponding to amino acids 33-55 at N-terminus of rat Slc12a1.
Purification Affinity purification
Storage buffer PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
Storage temperature Store at 4 °C. Aliquot to avoid repeated freezing and thawing.

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty