Anti-SLC12A1 Rabbit Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOPAB29036EA 636 GBP
ABNOPAB29036
Anti-SLC12A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Slc12a1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SLC12A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR