Anti-SLC9A3 Rabbit Polyclonal Antibody (Streptavidin)

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOPAB29035EA 556 GBP
ABNOPAB29035
Anti-SLC9A3 Rabbit Polyclonal Antibody (Streptavidin)
Rabbit polyclonal antibody raised against synthetic peptide of Slc9a3.

Recommended Dilutions: Immunocytochemistry Western Blot: 1:1000

Type:
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: Streptavidin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR