Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RCOR1 Mouse Monoclonal Antibody (ATTO 390) [clone: S72-8]

Supplier: Abnova
Mouse monoclonal antibody raised against partial recombinant RCOR1.

Recommended Dilutions: Western Blot: 1 μg/ml; Immunocytochemistry: 0.1-1 μg/ml; Immunohistochemistry: 0.1-1 μg/ml

Type: Primary
Antigen: RCOR1
Clonality: Monoclonal
Clone: S72-8
Conjugation: ATTO 390
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 μg 100 µg ABNOMAB11927
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol RCOR1
Antigen name REST corepressor 1
Conjugation ATTO 390
Clonality Monoclonal
Antibody type Primary
Clone S72-8
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms RCOR, COREST, KIAA0071
Amino acid number 109 to 293
Gene ID 23186
Format Liquid
Cross adsorption No
Immunogen Recombinant fusion protein corresponding to amino acids of 109-293, including ELM2 and SANT domains, of human RCOR1.
Purification Protein G Purification
Storage buffer PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
Storage temperature Store at 4 °C. Aliquot to avoid repeated freezing and thawing.

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty