Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNTNAP2 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S67-25]

Supplier: Abnova
Mouse monoclonal antibody raised against partial recombinant CNTNAP2.

Recommended Dilutions: Western Blot: 1 μg/ml; Immunocytochemistry: 0.1-1 μg/ml; Immunofluorescence: 1-10 μg/ml; Immunohistochemistry: 0.1-1 μg/ml

Type: Primary
Antigen: CNTNAP2
Clonality: Monoclonal
Clone: S67-25
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 μg 100 µg ABNOMAB11924
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CNTNAP2
Antigen name Contactin Associated Protein-like 2
Conjugation RPE (R-Phycoerythrin)
Clonality Monoclonal
Antibody type Primary
Clone S67-25
Host Mouse
Isotype IgG2a
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms AUTS15, NRXN4, CASPR2, CDFE, PTHSL1
Amino acid number 96 to 1265
Gene ID 26047
Format Liquid
Cross adsorption No
Immunogen Recombinant fusion protein corresponding to amino acids of 96-1265 at extracellular domain of human CNTNAP2.
Purification Protein G Purification
Storage buffer PBS, pH 7,4 (50% glycerol, 0,09% sodium azide)
Storage temperature Store at 4 °C. Aliquot to avoid repeated freezing and thawing.

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty