United Kingdom
Order Entry
Anti-HOXB2 Rabbit Polyclonal Antibody
  ABNOPAB25215
 :  Abnova
Anti-HOXB2 Rabbit Polyclonal Antibody
  ABNOPAB25215
 :  Abnova
 
£517.00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    homeobox B2
  • Antigen symbol:
    HOXB2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3212
  • Antigen synonyms:
    Hox-2.8|HOX2H|K8|HOX2
  • Storage buffer:
    PBS, pH 7,2 (0,05% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to HOXB2.
  • Purification:
    Affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against synthetic peptide of HOXB2.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: HOXB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human