Anti-C9ORF50 Rabbit Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOPAB21829EA 512 GBP
ABNOPAB21829
Anti-C9ORF50 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C9orf50.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: C9ORF50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR