Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDR Rabbit Polyclonal Antibody

Supplier: Abnova
Rabbit polyclonal antibody raised against synthetic peptide of KDR.

Recommended Dilutions: Western Blot: 0.1-1 μg/ml; ELISA: 0.01-0.1 μg/ml; Immunoprecipitation: 2-5 μg/ml; Immunohistochemistry: 2-5 μg/ml

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 μg 100 µg ABNOPAB12647
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol KDR
Antigen name Kinase Insert Domain Receptor
Reactivity Mouse, Dog, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Antigen synonyms Kinase insert domain receptor, Vascular endothelial growth factor receptor 2, FLK-1, FLK1, VEGFR-2, VEGFR2, Protein-tyrosine kinase receptor flk-1, CD309, Fetal liver kinase 1, KDR
Gene ID 3791
Format Liquid
Cross adsorption No
Immunogen A synthetic peptide corresponding to extracellular domain of human KDR.
Storage buffer PBS, pH 7,2 (10% Proclin300)
Storage temperature Store at 4 °C. For long term storage store at – 20 °C or lower. Aliquot to avoid repeated freezing and thawing.

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty