Anti-GFP Mouse Monoclonal Antibody [clone: GF28R]
ABNOMAB9749
: Abnova
- Antibody type:Primary
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Monoclonal
- Clone:GF28R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Jellyfish
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Storage buffer:10 mM PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to N-terminus of jellyfish GBP tag.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 Tests
Mouse monoclonal antibody raised against synthetic peptide of GFP tag.
Recommended Dilutions: Western Blot: 1:1000-1:3000; Immunohistochemistry: 1:500-1:2000
Type: Primary
Antigen: GFP
Clonality: Monoclonal
Clone: GF28R
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Jellyfishes