Anti-HDGFRP2 Rabbit Polyclonal Antibody

Supplier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ABNOPAB19389EA 512 GBP
ABNOPAB19389
Anti-HDGFRP2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of HDGF2.

Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: HDGFRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR