Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-ADGRF4 Rabbit Polyclonal Antibody

Supplier: Abnova
Rabbit polyclonal antibody raised against synthetic peptide of GPR115.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-10 μg/ml

Type: Primary
Antigen: ADGRF4
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Reactivity: Bovine, Gorilla, Horse, Human, Monkey, Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 μg 50 µg ABNOPAB26314
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol ADGRF4
Antigen name Adhesion G Protein-Coupled Receptor F4
Reactivity Monkey, Bovine, Gorilla, Horse, Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
ImmunoChemistry Yes
Antigen synonyms GPR115, PGR18
Gene ID 221393
Format Liquid
Cross adsorption No
Immunogen A synthetic peptide corresponding to 16 amino acids at C-terminus of human GPR115.
Purification Immunoaffinity chromatography
Storage buffer PBS (0,1% sodium azide)
Storage temperature Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty