Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPRIN2 Mouse Monoclonal Antibody

Supplier: Abnova
Mouse monoclonal antibody raised against a full-length recombinant GPRIN2.

Type: Primary
Antigen: GPRIN2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
Up to 5 Clones 1 ABNOH00009721-M
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol GPRIN2
Antigen name G Protein Regulated Inducer Of Neurite Outgrowth 2
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Host Mouse
Western blot Yes
ELISA Yes
Antigen synonyms KIAA0514, GRIN2
Amino acid number 1 to 461
Gene ID 9721
Format Liquid
Cross adsorption No
Immunogen GPRIN2 (AAH11672.1, 1 a.a. ~ 461 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Sequence MSSSHPEPGPWAPLSPRLQPLSQSSSSLLGEGREQRPELHKTASSTMWQAQLGEASTRPQAPEEEGNPPESMKPARASGPKARPSAGGHWRSSTVGNVSTMGGGDLCRLRAPSAAAMQRSHSDLVRSTQMRGHSGARKASLSCSALGSSPVHRAQLQPGGTSGQGGQAPAGLERDLAPEDETSNSAWMLGASQLSVPPLDLGDTTAHSSSAQAEPKAAEQLATTTCHALPPAALLCGMREMREVGAGGCCHALPATGILAFPKLVASVSESGLQAQHGVKIHCRLSGGLPGHSHCCAHLWGPAGLVPEPGSRTKDVWTMTSANDLAPAEASPLSAQDAGVQAAPVAACKAVATSPSLEAPAALHVFPEVTLGSSLEEAPSPVRDVRWDAEGMTWEVYGAAVDPEVLGVAIQKHLEMQFEQLQRAPASEDSLSVEGRRGPLRAVMQSLRRPSCCGCSGAAPE

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty