Anti-MYK Mouse Monoclonal Antibody (R-PE (Phycoerythrin) (R-Phycoerythrin)) [clone: 9E10]

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CAYM13426-1EA 495 GBP
CAYM13426-1
Anti-MYK Mouse Monoclonal Antibody (R-PE (Phycoerythrin) (R-Phycoerythrin)) [clone: 9E10]
Type: Tag
Antigen: MYK
Clonality: Monoclonal
Clone: 9E10
Conjugation: R-PE (Phycoerythrin) (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR