Safety workstation, Optiflow
Supplier: Bigneat
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIGNOF6003-FB04/03EA 2800 GBP
BIGNOF6003-FB04/03 BIGNOF6003-FB03/01 BIGNOF6003-FB01/03 BIGNOF6003-FB05/03 BIGNOF6003-FB02/03 BIGNOF6003-FB03/03
Safety workstation, Optiflow
Optiflow is a bench mounted ductless fume cabinet with a small footprint. It provides protection for the operator from chemical vapours, fumes or particles. It also provides unrestricted access for procedures and handlings that are difficult to perform within a standard fume cabinet. The down flow air takes dirty/contaminated air away from the operator and down through the perforations in the stainless steel work surface.
Optiflow is a bench mounted ductless fume cabinet with a small footprint. It provides protection for the operator from chemical vapours, fumes or particles. It also provides unrestricted access for procedures and handlings that are difficult to perform within a standard fume cabinet. The down flow air takes dirty/contaminated air away from the operator and down through the perforations in the stainless steel work surface.
  • Quiet operation
  • Keypad control panel with touch button controls
  • Visual and audible alarm sounds if airflow falls to an unacceptable level
  • Removable work surface for an easy cleaning
  • Access to the pre-filter, which protects the HEPA filter from larger particles and blockage from spills
  • Service due indicator
  • Simple to maintain
Housing and base made from epoxy coated mild steel, side panels from 8 mm tinted blue acrylic and work surface in stainless steel
  • Quiet operation
  • Keypad control panel with touch button controls
  • Visual and audible alarm sounds if airflow falls to an unacceptable level
  • Removable work surface for an easy cleaning
  • Access to the pre-filter, which protects the HEPA filter from larger particles and blockage from spills
  • Service due indicator
  • Simple to maintain
Select between 5 different models with carbon filter to protect against vapours or select the model with HEPA filter to eliminate particles with 0,3 microns or larger:
OS : Organic vapours
H+ : Acid vapours
F: Formaldehyde
MP : Multipurpose
AM : Ammonium
  • OS : Organic vapours
  • H+ : Acid vapours
  • F: Formaldehyde
  • MP : Multipurpose
  • AM : Ammonium