Sonic sifter separator
Supplier: Endecotts
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENDEZMSON-SON1EA 0 GBP
ENDEZMSON-SON1
Sonic sifter separator
The Sonic Sifter is a precision instrument for the rapid separation of a wide variety of dry particles and powders in the fine micron range. It will successfully separate samples down to 5 µ in as little as one minute, sometimes less, with consistent repeatability.
  • Simple and very quiet operation
  • Unique action
  • Very quick cycle time - typically less than one minute
  • Virtually no attrition of sample