Flow meter
Supplier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERC02901624EA 557 GBP
PERC02901624
Flow meter
Flow meter designed to measure the nitrogen flow.
  • Control and conserve purge gases
  • Includes 1/8 NPT female connection
Suitable for float displacement.