Buffered salt solutions, (PBS) Corning®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21-040-CMEA 108 GBP
21-040-CM 21-040-CV
Buffered salt solutions, (PBS) Corning®
Buffers Cell Culture Buffers
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate
  • Low endotoxin - <0,25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.