Anti-WBP1 Goat Polyclonal Antibody
USBIW9049-49
- Antibody type:Primary
- Antigen name:WW domain binding protein 1
- Antigen symbol:WBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:165 to 174
- Storage buffer:Tris saline buffer, pH 7,3, 0,5% BSA, 0,02% sodium azide.
- Sequence:C-PAHFEGTNVE
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Immunochemistry: 3. 75-7. 5 ?g/ml
Type: Primary
Antigen: WBP1 (WW domain binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human