Anti-STYX Rabbit Polyclonal Antibody
USBIS7985-50
- Antibody type:Primary
- Antigen name:serine/threonine/tyrosine interacting Protein
- Antigen symbol:STYX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Amino acid number:1 to 223
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: STYX (serine/threonine/tyrosine interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human