|
|
ORDER
SPECIFICATIONS
Antigen symbol | TRIM21 |
Antigen name | tripartite motif containing 21 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Goat |
Western blot | Yes |
ELISA | Yes |
Concentration | ~0,5 mg/ml |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA. |
Storage temperature | Cool |
Shipping temperature | Ice |
WRITE A REVIEW
REVIEWS (0)
Sort all by: