Anti-SRMS Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIS6567-60A-APEA 799 GBP
USBIS6567-60A-AP
Anti-SRMS Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: SRMS (src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR