|
|
ORDER
SPECIFICATIONS
Antigen symbol | SREBF1 |
Antigen name | Sterol regulatory element binding transcription factor 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody type | Primary |
Host | Rabbit |
Isotype | IgG |
Western blot | Yes |
ELISA | Yes |
Epitope | internal |
Purification | purified by immunoaffinity chromatography |
Form | liquid |
Storage buffer | PBS, 0,09% sodium azide. |
Storage temperature | Cool |
Shipping temperature | Ice |
WRITE A REVIEW
REVIEWS (0)
Sort all by: