Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SRC Mouse monoclonal antibody unconjugated [clone: 6D775]

Supplier: US Biological
Recommended dilution: Immunochemistry; Western Blot: 1:2000

Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: Monoclonal
Clone: 6D775
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µl 100 µl USBIS6505-02H
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SRC
Antigen name v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog
Reactivity Human, Monkey, Mouse
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 6D775
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoPrecipitation Yes
Amino acid number 477 to 947
Concentration 1 mg/ml
Purification purified
Form liquid
Storage buffer PBS containing 1mg/ml BSA and 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty