Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRNP Rabbit polyclonal antibody unconjugated

Supplier: US Biological
Recommended dilution: Western Blot: 0.5-2μg/ml

Type: Primary
Antigen: PRNP (prion protein)
Clonality: polyclonal
Conjugation: unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Sheep

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBIP8950-25F
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol PRNP
Antigen name Prion Protein
Reactivity Sheep, Bovine
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Amino acid number 142 to 148
Concentration ~0,5 mg/ml
Purification purified by Protein G chromatography
Form liquid
Storage buffer PBS, 0,2% gelatin, 0,05% sodium azide.
Storage temperature Cool
Shipping temperature Ice


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web. will publish your name as user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty