Anti-Gliadin Mouse Monoclonal Antibody [clone: 6F112]

Supplier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIG2035-85EA 689 GBP
USBIG2035-85
Anti-Gliadin Mouse Monoclonal Antibody [clone: 6F112]
Antibodies
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Gliadin
Clonality: monoclonal
Clone: 6F112
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR