United Kingdom
Order Entry
Anti-DGKD Rabbit polyclonal antibody
Anti-DGKD Rabbit polyclonal antibody
  USBID3441-84
undefined
Anti-DGKD Rabbit polyclonal antibody
  USBID3441-84
 
£626.00
   
  1 * 200 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    diacylglycerol kinase, delta 130kDa
  • Antigen symbol:
    DGKD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    C-terminal
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: DGKD (diacylglycerol kinase, delta 130kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse