Anti-COX 2, Human Mouse Monoclonal Antibody [clone: 4k1]
USBIC7904-84G-100
- Antibody type:Primary
- Antigen symbol:COX 2, Human
- Clonality:Monoclonal
- Clone:4k1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide before the addition of glycerol to 40%.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein A/G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1-10 ?g/ml; ELISA: 1:5000-1:50000
Type: Primary
Antigen: COX 2, Human
Clonality: Monoclonal
Clone: 4k1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse