Anti-CD68 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A326]
USBIC2423-05F
- Antibody type:Primary
- Antigen name:CD68 molecule
- Antigen symbol:CD68
- Clonality:Monoclonal
- Clone:6A326
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,4, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: Monoclonal
Clone: 6A326
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse